SL327 ≥99% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
α-[amino[(4-aminophenyl)thio]methylene]-2-(trifluoromethyl)-benzeneacetonitrile, (Z)-3-amino-3-(4-aminophenyl)sulphanyl-2-[2-(trifluoromethyl)phenyl]prop-2-enenitrile, 3-Amino-3-((4-aminophenyl)thio)-2-(2-(trifluoromethyl)phenyl)acrylonitrile
BMLEI3650010
ENZOBMLEI3650010EA
255
EUR
ENZOBMLEI3650010
SL327 ≥99% (durch HPLC)
SL327
A selective inhibitor of MEK1 and MEK2 (IC50= 0.18 and 0.22 µM respectively). Displays neuroprotective effects against ischemic brain injury.
- MEK inhibitor
White to off-white solid.
Soluble in DMSO to 100 mM or in ethanol to 100 mM.
Formel:
C₁₆H₁₂F₃N₃S Molecular Weight: 335,4 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD06411432 CAS-Nummer: 305350-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...