Dorsomorphin ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
6-(4-(2-(Piperidin-1-yl)ethoxy)phenyl)-3-(pyridin-4-yl)pyrazolo[1,5-a]pyrimidine, 6-[4-(2-Piperidin-1-ylethoxy)phenyl]-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine
Cell-permeable pyrrazolopyrimidine derivative that inhibits AMP kinase (Ki=109nM in the absence of AMP) in an ATP-competitive manner. It displays no significant inhibition of ZAPK, SYK, PKCT, PKA and JAK3. Decreases food intake in mice and inhibits the effects of AICAR and metformin. It has been shown to inhibit BMP type I receptors ALK2, ALK3 and ALK6. Promotes cardiomyogenesis in mouse embryonic stem cells. Induces protective autophagy in cancer cell lines.
- AMPK inhibitor
Yellow solid.
Soluble in DMSO (3mg/ml, warm)
Formel:
C₂₄H₂₅N₅O Molecular Weight: 399.5 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD08705402 CAS-Nummer: 866405-64-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...