SD-169 ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: p38 MAP Kinase Inhibitor VII, 1H-indole-5-carboxamide, Indole-5-carboxamide

BMLEI3710010
ENZOBMLEI3710010EA 86.9 EUR
ENZOBMLEI3710010
SD-169 ≥98%
SD-169

A selective inhibitor of p38α (IC50 = 3.2 nm) and p38β (IC50 = 3.2 nm), but not p38γ (IC50 > 50 μm), ERK2, JNK1 or MAPKAP2. Lowers blood glucose and improves glucose homeostasis in nonobese diabetic mice. Enhances axonal regeneration in animals with nerve transection.

Formel: C₉H₈N₂O
Molecular Weight: 160,17 g/mol
MDL: MFCD07779486
CAS-Nummer: 1670-87-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=792b56b08d1126eec8504825bb19f344d36e7d6d&vid=0223d268dc453c8245ac5f0cc118d2ab4cc2cf36&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR