(D-Pro1,2)-Retro-(D-HIV-Tat(48-57))
Lieferant: ENZO LIFE SCIENCES
BMLEI3840001
ENZOBMLEI3840001EA 391 EUR
ENZOBMLEI3840001
(D-Pro1,2)-Retro-(D-HIV-Tat(48-57))
Proteine und Peptide
Jnk inhibitor
Useful as a control peptide in studies using D-JNKi1 (BML-EI355). This peptide is the cell transporter, and does not contain the JIP (JNK inhibitor peptide) sequence.