AUDA ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
12-(3-Adamantan-1-yl-ureido)-dodecanoic acid, 12-(3-(Adamantan-1-yl)ureido)dodecanoic acid
BMLEI3920050
ENZOBMLEI3920050EA
348
EUR
ENZOBMLEI3920050
AUDA ≥98% (durch TLC)
AUDA
Novel inhibitor of soluble epoxide hydrolase (IC50=18nM and 69nM for the mouse and human enzymes respectively). Produces a hypotensive effect in angiotensin-infused rats and prevents and reverses cardiac hypertrophy in mouse models of cardiac hypertrophy.
- Epoxide hydrolase inhibitor
White solid.
Soluble in DMSO (10mg/ml) or 100% ethanol (4mg/ml).
Formel:
C₂₁H₃₆N₂O₃ Molecular Weight: 392,575 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD12912267 CAS-Nummer: 479413-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...