CCG-1423 ≥97% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-((1-((4-Chlorophenyl)amino)-1-oxopropan-2-yl)oxy)-3,5-bis(trifluoromethyl)benzamide, N-(2-(4-Chloroanilino)-1-methyl-2-oxoethoxy)-3,5-bis(trifluoromethyl)benzamide

BMLEI3940010
ENZOBMLEI3940010EA 86.9 EUR
ENZOBMLEI3940010
CCG-1423 ≥97% (durch HPLC)
CCG-1423

Cell permeable inhibitor of RhoA- and RhoC-mediated signaling. Inhibits LPA-induced DNA synthesis in PC-3 prostate cancer cell lines (<1µM) and inhibits the growth of RhoC-overexpressing melanoma cell lines (A375M2 and SK-Mel-147) at nanomolar concentrations.


  • Rho inhibitor


White solid.


Soluble in DMSO (10mg/ml) or 100% ethanol (9mg/ml).

Formel: C18H13ClF6N2O3
Molecular Weight: 454,8 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01566719
CAS-Nummer: 285986-88-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=267a7678fc270efc35cb92f2deb3c73c67336e8a&vid=83b2b4d789201de0e015311c69e7552735eff36a&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR