4,5,6,7-Tetrachloro-1H-indene-1,3(2H)-dione ≥95% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: TCID

BMLEI3990010
ENZOBMLEI3990010EA 132 EUR
ENZOBMLEI3990010
4,5,6,7-Tetrachloro-1H-indene-1,3(2H)-dione ≥95% (durch TLC)
4,5,6,7-Tetrachloro-1H-indene-1,3(2H)-dione

Novel inhibitor of ubiquitin C-terminal hydrolase L3 (IC50 = 0.6µM). Displays 125-fold greater selectivity over UCH-L1 (IC50 = 75µM).


  • UCH-L3 inhibitor


Orange solid.


Soluble in DMSO (25mg/ml).

Formel: C₉H₂Cl₄O₂
Molecular Weight: 283,93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00239209
CAS-Nummer: 30675-13-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ac741ae346d3115651d15a47af6fcc25772a790&vid=19b9ce87f63cceecfe47d31b7c1ff1ae97e63f98&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR