BML-282
Lieferant: ENZO LIFE SCIENCES
BMLEI4000005
ENZOBMLEI4000005EA 161 EUR
ENZOBMLEI4000005
BML-282
BML-282
UCH inhibitor
Novel inhibitor of ubiquitin C-terminal hydrolase L1 (IC50 = 0.81 µM). It displays 21-fold greater selectivity over UCH-L3 (IC50 = 17 µM).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d9bc47710627f2d869f248456ff5c7a27cbbb19&vid=28f6bafa7bff460f7a6f1dae2751c93f4e4c0dcc&prid=web&clid=SAPDAM