BML-282
Lieferant: ENZO LIFE SCIENCES
BMLEI4000005
ENZOBMLEI4000005EA
161
EUR
ENZOBMLEI4000005
BML-282
BML-282
UCH inhibitor
Novel inhibitor of ubiquitin C-terminal hydrolase L1 (IC50 = 0.81 µM). It displays 21-fold greater selectivity over UCH-L3 (IC50 = 17 µM).