BML-283 ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-ylsulphonyl)-4-(4-methoxyphenylsulphonyl)-piperazine
Pyruvate kinase M2 activator.
Potent (AC50=0.111 µM) and selective (over PKM1, PKR and PKL) activator of the tumor cell specific M2 isoform of pyruvate kinase (PKM2). Pyruvate kinase is a potential target for cancer chemotherapy. A useful new tool for altering aberrant glucose metabolism in cancer cells (Warburg effect).
Soluble in DMSO (12 mg/ml).
Bitte beachten Use/Stability: Store, as supplied, at room temperature for up to one year. Store solutions at -20°C for up to 3 months.
Formel:
C₁₉H₂₂N₂O₇S₂ Molecular Weight: 454,5 g/mol Schmelzpunkt: 229…231 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
562867-96-3 |
Spezifikation Testergebnisse
Purity | ≥98% (TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...