PAP-AMC, Tyrosinase substrate ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 1-(4-Methyl-coumarin-7-yl)-3-(4-hydroxyphenyl)urea

BMLEI4050100
ENZOBMLEI4050100EA 330 EUR
ENZOBMLEI4050100
PAP-AMC, Tyrosinase substrate ≥98% (durch TLC)
PAP-AMC, Tyrosinase substrate

Fluorogenic substrate for tyrosinase.


When assaying in phosphate buffer pH 7,1, the optimal excitation/emission wavelengths are 340/440 nm. 325/460 nm also work, but 380/460 nm will not detect this substrate.


Soluble in DMSO (18 mg/ml) or in methanol.


Bitte beachten Stable for at least 6 months after receipt when stored at –20 °C.

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR