Mg²⁺/ATP Activating Solution
Lieferant: ENZO LIFE SCIENCES
This solution contains pre-coupled Mg-ATP for immediate use as an energy source for a range of ATP-dependent reactions (e.g. ubiquitin thioester conjugation, proteasomal degradation, SUMO conjugation).
Liquid. In 50mM TRIS-Cl, pH 7,5, containing 0,1 M ATP and 0,1 M MgCl₂.