α-Linolensäure ≥99% (durch GC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 9Z,12Z,15Z-Octadecatriensäure, Linolensäure, cis,cis,cis-9,12,15-Octadecatriensäure, ALA

BMLFA0051000
ENZOBMLFA0051000EA 165 EUR
ENZOBMLFA0051000
α-Linolensäure ≥99% (durch GC)
α-Linolensäure

PPAR ligand.


  • Used for inducing adipocyte differentiation or for activating PPARs in cultured cells
  • Non-sterile


Useful for inducing adipocyte differentiation and/or activating PPARs in cultured cells, e.g. 200 and 250μM linolenic acid for, respectively, stimulation of FA transporter transcription in FAAR (NUC I) transfected 3T3-C2 cells and differentiation of Ob1771 preadipocytes.


Soluble in ethanol, hexane and methylene chloride.

Formel: C₁₈H₃₀O₂
Molecular Weight: 278.44 g/mol
Siedepunkt: 443
Schmelzpunkt: -9,5
MDL: MFCD00065720
CAS-Nummer: 463-40-1
Merck Index: 12,05530
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c56500549a753e8802f1015b3bedf65f31228b19&vid=a5bedb2ba1a7923f0f15a2bfb66eeaf678706a8a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥99% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR