Docosa-7Z,10Z,13Z,16Z,19Z-pentaenoic acid 98

Lieferant: ENZO LIFE SCIENCES

Synonyme: (7Z,10Z,13Z,16Z,19Z)-docosa-7,10,13,16,19-pentaenoic acid, Clupanodonic acid

BMLFA0120010
ENZOBMLFA0120010EA 84.7 EUR
ENZOBMLFA0120010
Docosa-7Z,10Z,13Z,16Z,19Z-pentaenoic acid 98
Docosa-7Z,10Z,13Z,16Z,19Z-pentaenoic acid

Polyunsaturated fatty acid.


Soluble in ethanol, hexane and methylene chloride.

Formel: C₂₂H₃₄O₂
Molecular Weight: 330.512 g/mol
CAS-Nummer: 24880-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ca07de3038367e4f2e5a923370fe1125481c3a2&vid=7cbe064a6dd25f7a21f1e1647fccae116652eabc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98%
Appearance Clear to pale yellow oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR