Linoleamide ≥98% (durch TLC), weißer Feststoff

Lieferant: ENZO LIFE SCIENCES

BMLFA0230010 BMLFA0230050
ENZOBMLFA0230010EA 66 EUR
ENZOBMLFA0230010 ENZOBMLFA0230050
Linoleamide ≥98% (durch TLC), weißer Feststoff
Linoleamide

Unsaturated analog of endogenous sleep inducing lipid.


Soluble in 100% ethanol or in DMSO.

Formel: C₁₈H₃₃NO
MDL: MFCD00798239
CAS-Nummer: 3072-13-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f845a75711e1331b25bf57ccf753cfb9c06637e&vid=10c0afa086bbcef33061ea3c0d6869ffee6be9bc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR