Eicosa-5Z,8Z-dienoic acid ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLFA0240001
ENZOBMLFA0240001EA 180 EUR
ENZOBMLFA0240001
Eicosa-5Z,8Z-dienoic acid ≥98% (durch TLC)
Eicosa-5Z,8Z-dienoic acid

Arachidonic acid analog.


Arachidonic acid analog lacking the C-11 and C-14 double bonds.


Soluble in ethanol or DMSO.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f612710e6a3cbdd2a61fad0f39dc2fb52b35a2f&vid=b645791030c8711330dd85a91925d884c9f87ac7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR