N-Arachidonylglycine ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: NAGly

BMLFA0290005
ENZOBMLFA0290005EA 58.3 EUR
ENZOBMLFA0290005
N-Arachidonylglycine ≥98% (durch TLC)
N-Arachidonylglycine

FAAH inhibitor


Endogenous anandamide-like compound. Lacks affinity for CB1 receptors (Ki>10µM), TRPV1 (EC50>10µM) and anandamide uptake (IC50>50µM), but inhibits fatty acid amide hydrolase (FAAH) (IC50=8.5µM-50µM, depending on cell type and species). Displays anti-inflammatory and analgesic activity. Identified as an insulin secretagogue in a primary beta-cell-based functional assay. Represents a new class of "lipoamino acids".

Formel: C₂₂H₃₅NO₃
Molecular Weight: 361,526 g/mol
MDL: MFCD03791313
CAS-Nummer: 179113-91-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=102a96392aa9eaa10c1bc7e336e85886e9a53da8&vid=5ec3232097426b69e407134d9bed89627ef05b7c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR