Virodhamine ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: 2-Aminoethyl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate

BMLFA0360005
ENZOBMLFA0360005EA 184 EUR
ENZOBMLFA0360005
Virodhamine ≥98% (durch TLC)
Virodhamine

Cannabinoid receptor ligand.


Endogenous cannabinoid which has been found to occur at concentrations 2 to 9 fold greater than anandamide in peripheral tissue. It acts as a full agonist at the CB₂ receptor and an antagonist at the CB₁ receptor.


Soluble in DMSO (45 mg/ml), dimethyl formamide (40 mg/ml) or ethanol (20 mg/ml).

Formel: C₂₂H₃₇NO₂
Molecular Weight: 347,53 g/mol
MDL: MFCD06654562
CAS-Nummer: 443129-35-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56e7946985f4d80308a40ab2148420e09611f998&vid=ce67a9bbd22a07d156f9704fd69d8dd0faf68474&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR