N-Arachidonoyl-L-phenylalanine ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-(5Z,8Z,11Z,14Z-icosatetraenoyl)-L-phenylalanine, L-Phenylalanine, N-[(5Z,8Z,11Z,14Z)-1-oxo-5,8,11,14-eicosatetraen-1-yl]-, N-arachidonoylphenylalanine

BMLFA0400025
ENZOBMLFA0400025EA 384 EUR
ENZOBMLFA0400025
N-Arachidonoyl-L-phenylalanine ≥98% (durch TLC)
N-Arachidonoyl-L-phenylalanine

Putative endogenous member of a growing family of lipo-amino acids (N-acyl amino acids).


Soluble in DMSO (>25 mg/ml) or 100% ethanol (>25 mg/ml).


Bitte beachten Subject to oxidation.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb53a789157f1750aed7131e3c5060134661b90e&vid=8db47c0e6cc6ab1d092988b4f23aeb4553181ab0&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR