21-[4-(2,6-Di-1-pyrrolidinyl-4-pyrimidinyl)-1-piperazinyl]pregna-1,4,9[11]triene-3,20-dione maleate salt ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
BMLFR1000100
ENZOBMLFR1000100EA
161
EUR
ENZOBMLFR1000100
21-[4-(2,6-Di-1-pyrrolidinyl-4-pyrimidinyl)-1-piperazinyl]pregna-1,4,9[11]triene-3,20-dione maleate salt ≥98% (durch TLC)
21-[4-(2,6-Di-1-pyrrolidinyl-4-pyrimidinyl)-1-piperazinyl]pregna-1,4,9[11]triene-3,20-dione maleate salt
Blocks lipid peroxidation
Lazaroid inhibitor of iron-dependent lipid peroxidation. Antioxidant. Inhibits lipid peroxidation in a variety of experimental systems including cultured brain microvessel endothlial cells, Cu2+ treated monocyclic THP1 cells and LLC-PK1 cell layers where it blocks F2 isoprostane production. Prevented organ damage in intestinal cold storage, preservation and reperfusion injury.
Formel:
C₄₁H₅₄N₆O₆ |
MDL:
MFCD01074991 CAS-Nummer: 153190-29-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...