L-Buthionine sulphoximine ≥97% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: L-Buthionine-(S,R)-sulphoximine, (2S)-2-Amino-4-(butylsulphonimidoyl)butanoic acid

BMLFR1170500
ENZOBMLFR1170500EA 215 EUR
ENZOBMLFR1170500
L-Buthionine sulphoximine ≥97% (durch TLC)
L-Buthionine sulphoximine

L-Buthionine-sulfoximine (BSO) induces oxidative stress in cells by irreversibly inhibiting γ-glutamylcysteine synthetase, an essential enzyme for the synthesis of glutathione.


  • γ-Glutamyl transpeptidase inhibitor


White solid.


Soluble in water.

Formel: C₈H₁₈N₂O₃S
Molecular Weight: 222,303 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067000
CAS-Nummer: 83730-53-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6e83849b4c87d24829e2b15268370f196a49bf6&vid=3efd40665a75b8f0b12bf9ea687747d9c6806722&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR