Triacetyl resveratrol ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
Synonyme:
(E)-5-(4-Acetoxystyryl)-1,3-phenylene diacetate, 3,5,4′-Tri-O-acetylresveratrol, Acetyl-trans-resveratrol, 5-[(1E)-2-[4-(acetyloxy)phenyl]ethenyl]-1,3-Benzenediol-1,3-diacetate
BMLFR1190010
ENZOBMLFR1190010EA
84.7
EUR
ENZOBMLFR1190010
Triacetyl resveratrol ≥98% (durch TLC)
Triacetyl resveratrol
SIRT activator.
A resveratrol prodrug. One approach to increasing the half life of resveratrol in vivo is acetylation of the phenolic OH groups. Deacetylation presumably takes place in vivo or in intact cells via the action of intracellular esterases.
Soluble in DMSO (25 mg/ml) or 100% ethanol (warm) (8 mg/ml).
Formel:
C₂₀H₁₈O₆ Molecular Weight: 354.35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01546481 CAS-Nummer: 42206-94-0 |
|
Spezifikation Testergebnisse
Purity | ≥98% (TLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...