Bordetella pertussis toxin
Lieferant: ENZO LIFE SCIENCES
BMLG1010050
ENZOBMLG1010050EA
431
EUR
ENZOBMLG1010050
Bordetella pertussis toxin
Mikrobiologische Testsysteme
Antigen-Antikörper-Tests
Ag-AK, Seren
Bordetella pertussis toxin (Islet-activating protein, PTX) from Bordetella pertussis, strain NCTC 10739. is a high purity toxin applicable to ELISA, antibody capture, cytotoxicity studies, LINE assay.
- High purity, native protein available as >98% pure
- Minimal lot-to-lot variation ensures reproducible results
- Lyophilised format for extended shelf-life