Clostridium difficile toxin A
Lieferant: ENZO LIFE SCIENCES
BMLG1400050
ENZOBMLG1400050EA
478
EUR
ENZOBMLG1400050
Clostridium difficile toxin A
Mikrobiologische Testsysteme
Antigen-Antikörper-Tests
Ag-AK, Seren
Clostridium difficile toxin A from Clostridium difficile, strain VPI10463 is a high potency toxin applicable to ELISA, antibody capture, cytotoxicity studies and LINE assay.
- Minimal lot to lot variation
- Single lot bulk quantities available
- Lyophilised for ease of handling and long term stability