Clostridium difficile toxoid A
Lieferant: ENZO LIFE SCIENCES
BMLG1450050
ENZOBMLG1450050EA
517
EUR
ENZOBMLG1450050
Clostridium difficile toxoid A
Mikrobiologische Testsysteme
Antigen-Antikörper-Tests
Ag-AK, Seren
Clostridium difficile toxoid A from Clostridium difficile, strain VPI10463 is a high potency toxin applicable to ELISA, antibody capture, cytotoxicity studies, immunogen and LINE assay.
- Minimal lot-to-lot variation
- Inactivated by formaldehyde
- Lyophilised for ease of handling and long term stability