Clostridium difficile toxoid A
Lieferant: ENZO LIFE SCIENCES
BMLG1450050
ENZOBMLG1450050EA 517 EUR
ENZOBMLG1450050
Clostridium difficile toxoid A
Mikrobiologische Testsysteme Antigen-Antikörper-Tests Ag-AK, Seren
Clostridium difficile toxoid A from Clostridium difficile, strain VPI10463 is a high potency toxin applicable to ELISA, antibody capture, cytotoxicity studies, immunogen and LINE assay.
  • Minimal lot-to-lot variation
  • Inactivated by formaldehyde
  • Lyophilised for ease of handling and long term stability