Ac-Cys(farnesyl)-OH ≥98% (durch TLC)

Lieferant: Enzo Life Sciences

BMLG2000005 BMLG2000025
ENZOBMLG2000005EA 86.9 EUR
ENZOBMLG2000005 ENZOBMLG2000025
Ac-Cys(farnesyl)-OH ≥98% (durch TLC)
Ac-Cys(farnesyl)-OH

Methyltransferase inhibitor


Members of the Ras superfamily of G proteins, gamma-subunits of heterotrimeric G proteins and the alpha-subunit of cGMP phosphodiesterase from retinal rods are all carboxyl methylated at carboxy-terminal S-farnesylcysteine residues. AFC specifically inhibits the S-farnesylcysteine methyl transferase (Km=20 µM) in cell free extracts and in whole cells. It inhibits carboxyl methylation of p21ras platelet Rap15 and the gamma-subunit of transducin. It blocks receptor-mediated signal transduction in platelets and neutrophils not by inhibiting the methyltransferase, but possibly by inhibiting capacitative Ca2+ entry.

Formel: C₂₀H₃₃NO₃S
MDL: MFCD00871540
CAS-Nummer: 135304-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9c82c2e9a8c14be8f775b349968828d02b575b2&vid=85a872348fc8793b0b82adc493a409199d85a9d3&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR