Mevastatin sodium ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Mevastatin sodium salt
BMLG2380005
ENZOBMLG2380005EA
114
EUR
ENZOBMLG2380005
Mevastatin sodium ≥98% (durch HPLC)
Mevastatin sodium
This is the active carboxylate form of mevastatin. It is active in whole cells as well as in cell free assays.
- HMG-CoA reductase inhibitor
White to off-white solid.
Soluble in ethanol (20 mg/ml) and DMSO (30 mg/ml).
Formel:
C₂₃H₃₅O₆·Na Molecular Weight: 430,51 g/mol Schmelzpunkt: 44…45 °C Lagertemperatur: Gefrierschrank |
CAS-Nummer:
99782-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...