Lovastatin hydroxy acid sodium ≥97% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (3R,5R)-7-[(1S,2S,6R,8S,8aR)-2,6-dimethyl-8-[[(2S)-2-methylbutanoyl]oxy]-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic acid sodium salt, Lovastatin sodium salt, 6-α-Methylcompactin sodium salt, Lovastatin EP Impurity B Sodium Salt, Lovastatin EP Impurity B, Sodium (3R,5R)-7-((1S,2S,6R,8S,8aR)-2,6-dimethyl-8-(((S)-2-methylbutanoyl)oxy)-1,2,6,7,8,8a-hexahydronaphthalen-1-yl)-3,5-dihydroxyheptanoate, Mevinolin sodium salt

BMLG2450005
ENZOBMLG2450005EA 86.9 EUR
ENZOBMLG2450005
Lovastatin hydroxy acid sodium ≥97% (durch HPLC)
Lovastatin hydroxy acid sodium

Active carboxylate form of Lovastatin. Active in whole cells as well as cell-free assays. Inhibits cholesterol/ isoprenoid biosynthesis by inhibition of HMG-CoA reductase (Ki for acid form is 1 nM) and blocks protein isoprenylation and reduces plasma cholesterol levels in humans. Causes cells to arrest early in the G1 phase.


  • Active in whole cells as well as cell-free assays


White solid.


Soluble in DMSO (10mg/ml) or 100% ethanol (10mg/ml).

Formel: C₂₄H₃₇O₆·Na
Molecular Weight: 444,54 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00870475
CAS-Nummer: 75225-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR