TTNPB ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
Synonyme:
Arotinoid acid, (E)-4-[2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)-1-propenyl]benzoic acid, (E)-4-(2-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)prop-1-en-1-yl)benzoic acid, TTNPB (Arotinoid Acid)
BMLGR1050005
ENZOBMLGR1050005EA
135
EUR
ENZOBMLGR1050005
TTNPB ≥98% (durch HPLC)
TTNPB
Retinoic acid receptor ligand.
Retinoic acid analog which acts as a selective RAR agonist.
Soluble in DMSO or ethanol (with gentle warming).
Formel:
C₂₄H₂₈O₂ Molecular Weight: 348,5 g/mol |
MDL:
MFCD00673917 CAS-Nummer: 71441-28-6 |
Spezifikation Testergebnisse
Purity | ≥98% (HPLC) |
Identity | Determined by H-NMR |
Appearance | White crystalline solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...