Adapalene ≥98%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic Acid, 6-(3-(Adamantan-1-yl)-4-methoxyphenyl)-2-naphthoic acid
BMLGR1110020
ENZOBMLGR1110020EA
135
EUR
ENZOBMLGR1110020
Adapalene ≥98%
Adapalene
Retinoic acid receptor ligand.
Adapalene is a stable analog of retinoic acid and acts selectively as an agonist at RAR beta and RAR gamma. It inhibits proliferation and induces apoptosis in colorectal cancer cells. Its unique pharmacological properties make it superior to other retinoids for the treatment of acne.
Formel:
C₂₈H₂₈O₃ Molecular Weight: 412,53 g/mol |
MDL:
MFCD03106112 CAS-Nummer: 106685-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...