AC-55469 ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
4'-Octyl-[1,1'-biphenyl]-4-carboxylic acid, AC-55649
BMLGR1120010
ENZOBMLGR1120010EA
89.1
EUR
ENZOBMLGR1120010
AC-55469 ≥98% (durch HPLC)
AC-55469
AC-55649 is potent and subtype selective agonist at RARβ2 ( EC50=6.9 µM). It inhibits proliferation of MCF-7 cells and induces differentiation in a neuronal cell line.
- Retinoic acid receptor ligand
White to gray, Crystals or crystalline powder
Molecular Weight:
310,4 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00157040 CAS-Nummer: 59662-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...