({4-Chlor-6-[(2,3-dimethylphenyl)amino]-2-pyrimidinyl}sulfanyl)essigsäure ≥97% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Gefahr

Synonyme: 2-((4-Chloro-6-((2,3-dimethylphenyl)amino)pyrimidin-2-yl)thio)acetic acid, WY-14643, Pirinixic acid

BMLGR2000250
ENZOBMLGR2000250EA 487 EUR
ENZOBMLGR2000250
({4-Chlor-6-[(2,3-dimethylphenyl)amino]-2-pyrimidinyl}sulfanyl)essigsäure ≥97% (durch HPLC)
[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid

PPAR ligand.


WY-14643 is a potent and widely used peroxisome proliferator activator receptor (PPAR) activator and act as a selective for PPARα.


Soluble in ethyl acetate or dilute base or slightly soluble in alcohol.

Formel: C₁₄H₁₄ClN₃O₂S
Molecular Weight: 323.80 g/mol
Schmelzpunkt: 155…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191335
CAS-Nummer: 50892-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16cbf8b3665c27b778a9e33fb85e27d79d7be04d&vid=c52b377d9a32ef5464f0e48f0b950593a91330e6&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥97% (HPLC)
Identity Determined by NMR
Appearance White to off-white solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR