6-Formylindolo[3,2-b]carbazole ≥95% (durch HPLC), gelber Feststoff
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Synonyme:
FICZ, 5,11-Dihydroindolo[3,2-b]carbazole-6-carbaldehyde
BMLGR2060100
ENZOBMLGR2060100EA
165
EUR
ENZOBMLGR2060100
6-Formylindolo[3,2-b]carbazole ≥95% (durch HPLC), gelber Feststoff
6-Formylindolo[3,2-b]carbazole
Ah receptor ligand.
A tryptophan-derived, high affinity aryl hydrocarbon receptor ligand (Kd=7×10-11 M). Proposed to be an endogenous AhR ligand. Induces rapid and transient expression of cytochrome P450-1A1 (CYP1A1) in human cells at 100 pM.
Soluble in DMSO (10 mg/ml).
Formel:
C₁₉H₁₂N₂O Molecular Weight: 284,32 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD09879259 CAS-Nummer: 172922-91-7 |
|
Spezifikation Testergebnisse
Purity | ≥95% (HPLC) |
Appearance | Yellow solid |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...