3,3'-Methylenediindole ≥98% (durch HPLC), weißgraues Puder

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: DIM, 3,3'-Diindolylmethane

BMLGR2070250
ENZOBMLGR2070250EA 361 EUR
ENZOBMLGR2070250
3,3'-Methylenediindole ≥98% (durch HPLC), weißgraues Puder
3,3'-Methylenediindole

Ah receptor antagonist.


A partial antagonist for the aryl hydrocarbon receptor. Induces apoptosis in human cancer cells. Activates cytochrome P-450 1A2.


Soluble in DMSO.

Formel: C₁₇H₁₄N₂
Molecular Weight: 246,31 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00195766
CAS-Nummer: 1968-05-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9cd4f2f49aaf34c38192a73b51c59060e22432f&vid=1293df9ac089230f4ed07c71484b2cf5586a6e0b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR