TCPOBOP ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 1,4-Bis((3,5-dichloropyridin-2-yl)oxy)benzene

BMLGR2450025
ENZOBMLGR2450025EA 180 EUR
ENZOBMLGR2450025
TCPOBOP ≥98% (durch TLC)
TCPOBOP

Constitutive androstane receptor (CAR) activator.


TCPOBOP is an activator of the mouse constitutive androstane receptor (CAR) and a potent inducer of cytochrome P450 monoxygenase activity and also have a potent tumor-promoting activity and hepatocarcinogenic effects in mice but not in rats.


Soluble in DMSO (5 mg/ml).

Formel: C₁₆H₈Cl₄N₂O₂
Molecular Weight: 402,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00057368
CAS-Nummer: 76150-91-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9da8be96e7bd2650089ca5d00837755f30fa392c&vid=f3d269a9c76b95a70ea10d0d7408b9e1ca887c50&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Source Synthetic
Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR