WAY-200070 ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
Synonyme:
7-Bromo-2-(4-hydroxyphenyl)benzo[d]oxazol-5-ol
BMLGR2470010
ENZOBMLGR2470010EA
161
EUR
ENZOBMLGR2470010
WAY-200070 ≥98% (durch TLC)
WAY-200070
Estrogen receptor ligand.
Selective estrogen receptor ERβ agonist displaying 68-fold greater selectivity over ERα (EC50 = 2 and 155 nM, respectively). It stimulates galanin expression in GnRH neurons and displays anxiolytic/antidepressant activity in various in vivo rodent models (administration to mice at 30 mg/kg s.c.).
Soluble in DMSO (12 mg/ml) or 100% ethanol (2.5 mg/ml).
Formel:
C₁₃H₈BrNO₃ Molecular Weight: 306,115 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16618385 CAS-Nummer: 440122-66-7 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Purity | ≥98% (TLC) |
Appearance | Off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...