2,3-Bis(4-hydroxyphenyl)propannitril ≥98% (durch TLC), weißer Feststoff
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Synonyme:
DPN
BMLGR2480050
ENZOBMLGR2480050EA
558
EUR
ENZOBMLGR2480050
2,3-Bis(4-hydroxyphenyl)propannitril ≥98% (durch TLC), weißer Feststoff
2,3-Bis(4-hydroxyphenyl)propionitrile
Estrogen receptor β activator.
Selective estrogen receptor ERβ agonist displaying 70-fold greater selectivity over ERα (EC50 = 0.85 and 66 nM, respectively). It displays neuroprotective activity in various in vitro and in vivo models.
Soluble in DMSO (10 mg/ml) or 100% ethanol (1.5 mg/ml).
Formel:
C₁₅H₁₃NO₂ Lagertemperatur: Gefrierschrank |
MDL:
MFCD01695412 CAS-Nummer: 1428-67-7 |
|
Spezifikation Testergebnisse
Purity | ≥98% (TLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...