Coumermycin A1 ≥97% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLGR3170005
ENZOBMLGR3170005EA 182 EUR
ENZOBMLGR3170005
Coumermycin A1 ≥97% (durch TLC)
Coumermycin A1

Inhibits supercoiling activity of gyrase by binding to the gyrase B domain – but more importantly is a novel cell permeable dimerizing ligand for gyrase B fusion proteins. Has been used to dimerize a variety of signaling proteins such as Raf1, JAK2, Tyk and L-selectin.


  • White to light yellow solid


Soluble in DMSO

Formel: C₅₅H₅₉N₅O₂₀
Molecular Weight: 1110,1 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 4434-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR