Prima-1 ≥98% (durch TLC), weißer bis weißgrauer Feststoff

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: 2,2-Bis(hydroxymethyl)-3-quinuclidinone, 2,2-Bis(hydroxymethyl)quinuclidin-3-one

BMLGR3330005
ENZOBMLGR3330005EA 84.7 EUR
ENZOBMLGR3330005
Prima-1 ≥98% (durch TLC), weißer bis weißgrauer Feststoff
Prima-1

p53 activator.


Novel cell-permeable p53 reactivating agent. Restores sequence-specific DNA binding and the active conformation to mutant p53 (in 13 of 14 p53 mutants tested) in vitro and in living cells. Induces apoptosis in human tumor cell lines and displays antitumor activity in mice.


Soluble at 25 mg/ml in ethanol, 100% DMSO and DMF. Soluble at 10 mg/ml in water.


Bitte beachten Product is not sterile.

Formel: C₉H₁₅NO₃
Molecular Weight: 185.223 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD04974196
CAS-Nummer: 5608-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05f7864a444f31113dafbeede5ea2ab04c67b820&vid=f4535ec2ffc9f161f28a77c255dab09c7d0727a1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR