Tomatidine HCl ≥90% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLGR3350025
ENZOBMLGR3350025EA 67.1 EUR
ENZOBMLGR3350025
Tomatidine HCl ≥90% (durch TLC)
Tomatidine HCl

Negative control for cyclopamine.


Steroidal alkaloid structurally similar to cyclopamine but does not inhibit the hedgehog (shh) pathway. Used as a negative control for jervine.


Soluble in DMSO (2 mg/ml), 100% ethanol (2.5 mg/ml) or dimethyl formamide (2 mg/ml).

Formel: C₂₇H₄₅NO₂·HCl
Lagertemperatur: Gefrierschrank
CAS-Nummer: 6192-62-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57258e16b1f2fe9bbf71975d62a807794da8530f&vid=bd9f6294b5d0bd034c048ab39cf7c2f9356404f0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Source Tomato leaves
Purity ≥90% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR