Zebularine ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
BMLGR3440010
ENZOBMLGR3440010EA
161
EUR
ENZOBMLGR3440010
Zebularine ≥98% (durch TLC)
Zebularine
Dnmt inhibitor
- Purification: TLC: 30% Methanol/methylene chloride + 1% H2O, Rf = 0.3
Zebularine is a DNA methylation inhibitor which acts by forming a covalent complex with DNMT. It displays antiangiogenic activity and markedly inhibits vessel formation in different tumor models. It reactivates silenced E-cadherin and potentiates the effects of other epigenetic therapeutics such as vorinistat. Induces differentiation of mesenchymal cells into cardiomyocytes. Inhibits cytidine deaminase.
Formel:
C₉H₁₂N₂O₅ |
MDL:
MFCD04973699 CAS-Nummer: 3690-10-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...