ITSA-1 ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (1H-Benzo[d][1,2,3]triazol-1-yl)(2,4-dichlorophenyl)methanone

BMLGR3500025
ENZOBMLGR3500025EA 114 EUR
ENZOBMLGR3500025
ITSA-1 ≥98% (durch TLC)
ITSA-1

HDAC inhibitor


ITSA-1 is a cell permeable benzotriazole amide that counteracts trichostatin A (Prod. No. BML-GR309) induced cell cycle arrest, histone acetylation and transcriptional activation but not the activity of other HDAC inhibitors.

Formel: C₁₃H₇Cl₂N₃O
Molecular Weight: 292,1202 g/mol
MDL: MFCD00195103
CAS-Nummer: 200626-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bc75d0839504c4383b0d150f368acf6c2fa7286&vid=4f1d8d40ccf2af50728ef93db1a8aeccef453b16&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR