Histone Deacetylase Inhibitor VIII ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: tert-Butyl (4-(3-((7-(hydroxyamino)-7-oxoheptyl)carbamoyl)isoxazol-5-yl)phenyl)carbamate, BML-281, CAY10603, HDAC6 Inhibitor

BMLGR3610001 BMLGR3610005
ENZOBMLGR3610001EA 114 EUR
ENZOBMLGR3610001 ENZOBMLGR3610005
Histone Deacetylase Inhibitor VIII ≥98% (durch TLC)
Histone Deacetylase Inhibitor VIII

HDAC6 inhibitor


Selective HDAC6 inhibitor (IC50=0.002nM). IC50 values for other HDACs are 271, 252, 0.42, 6851 and 90.7 nM for HDACs 1,2,3,8 and 10 respectively. Potent inhibitor of the proliferation of a variety of pancreatic cancer cell lines (IC50s in the 0.1-0.6µM range).

Formel: C₂₂H₃₀N₄O₆
MDL: MFCD17010286
CAS-Nummer: 1045792-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45565e7a1092844a24072993dae8e6b5cb79f66b&vid=efd3c03c8d163c520b2f60cb93ad0fa0c5e6cf40&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR