Tubacin ≥96%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
rel-N1-(4-((2R,4R,6S)-4-(((4,5-Diphenyloxazol-2-yl)thio)methyl)-6-(4-(hydroxymethyl)phenyl)-1,3-dioxan-2-yl)phenyl)-N8-hydroxyoctanediamide
BMLGR3620500
ENZOBMLGR3620500EA
527
EUR
ENZOBMLGR3620500
Tubacin ≥96%
Tubacin
HDAC inhibitor
Synthetic.
Tubacin (tubulin acetylation inducer) is a highly potent and selective, reversible, cell-permeable inhibitor of HDAC6 (IC50=0.004µM). IC50's for the other HDACs are 1000-fold higher, making tubacin both more selective and more potent than Tubastatin A, which also inhibits HDAC8. Concentration in cell culture experiments typically ranges from 2-50µM.
Formel:
C₄₁H₄₃N₃O₇S Molecular Weight: 721,86 g/mol |
MDL:
MFCD28144524 CAS-Nummer: 537049-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...