5(S)-Hydroxyeicosa-6E,8Z,11Z,14Z-tetraenoic acid (HETE) ≥98%

Lieferant: ENZO LIFE SCIENCES
Gefahr

BMLH0050050
ENZOBMLH0050050EA 324 EUR
ENZOBMLH0050050
5(S)-Hydroxyeicosa-6E,8Z,11Z,14Z-tetraenoic acid (HETE) ≥98%
5(S)-Hydroxyeicosa-6E,8Z,11Z,14Z-tetraenoic acid (HETE)

Biosynthesis via 5-lipoxygenase. Chemotactic and chemokinetic for PMNL. Induces degranulation and calcium flux.

Formel: C₂₀H₃₂O₃
Lagertemperatur: Ultra-Tiefkühlschrank (–80°C)
CAS-Nummer: 70608-72-9
UN: 1170
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9643aecf027fa0bd42c230296cd22f8ea61836c&vid=9ff15e72c5f3acdc7b3dfc402686ef48b8e20694&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Formulation Solution in ethanol.
Purity ≥98%
Concentration 50 µg/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR