8(S)-Hydroxyeicosa-5Z,9E,11Z,14Z-tetraenoic acid (HETE)-Lösung 50 µg/ml in Ethanol , 99% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
BMLH0080050
ENZOBMLH0080050EA
316
EUR
ENZOBMLH0080050
8(S)-Hydroxyeicosa-5Z,9E,11Z,14Z-tetraenoic acid (HETE)-Lösung 50 µg/ml in Ethanol , 99% (durch HPLC)
8(S)-Hydroxyeicosa-5Z,9E,11Z,14Z-tetraenoic acid (HETE)
Activator of PKC and PPARα.
Biosynthesis in human neutrophils, phorbol ester-treated mouse epidermis and by hepatic microsomal cytochrome P-450. Chemotactic for neutrophils. Activator of PPARα. Activates mouse skin protein kinase C (PKC).
Formel:
C₂₀H₃₂O₃ Lagertemperatur: Ultra-Tiefkühlschrank (–80°C) |
CAS-Nummer:
98462-03-4 UN: 1170 ADR: 3,II |
Spezifikation Testergebnisse
Concentration | 50 μg/ml |
Formulation | In ethanol |
Purity | ≥99% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...