12(S)-Hydroxy-(5Z,8Z,10E,14Z)-eicosatetraenoic acid 50 µg/ml in Ethanol ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
Biosynthesis by platelet 12-lipoxygenase. Chemotactic and chemokinetic for PMNL2 and for vascular smooth muscle cells. Antiproliferative effect on smooth muscle cells. Second messenger in angiotensin induced aldosterone production. Induces endothelial cell retraction and tumor cell adhesion.
Formel:
C₂₀H₃₂O₃ Lagertemperatur: Ultra-Tiefkühlschrank (–80°C) |
MDL:
MFCD00036916 CAS-Nummer: 54397-83-0 UN: 1170 ADR: 3,III |
|
Spezifikation Testergebnisse
Concentration | 50 µg/ml |
Formulation | Solution in ethanol |
Purity | ≥98% (HPLC) |
Identity | Determined by NMR |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...