13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid ≥99%

Lieferant: ENZO LIFE SCIENCES

Synonyme: S)-13-HODE

BMLHO0130050
ENZOBMLHO0130050EA 86.9 EUR
ENZOBMLHO0130050
13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid ≥99%
13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid

Inhibits thromboxane production in human platelets


Biosynthesis in rabbit peritoneal serous membranes, atherosclerotic plaques, healthy blood vessels, psoriatic skin, endothelial cells. Stimulates prostacyclin production by endothelial cells. Inhibits thromboxane production in human platelets. Inhibits platelet and tumor cell adhesion to endothelium. Antagonizes 12-HETE-induced tumor cell adhesion to endothelium. Stimulates DNA synthesis in BALB/c 3T3 fibroblasts.

Formel: C₁₈H₃₂O₃
Molecular Weight: 296,45 g/mol
MDL: MFCD00065833
CAS-Nummer: 29623-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ff65ea86dc3c0e131b33ef4d29d3945424c92f2&vid=3d3d3ebca2f53e168b8167a3b53f0d58eb2ec120&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR