12(S)-Hydroperoxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HPETE) ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 12(S)-HPETE

BMLHP0120050
ENZOBMLHP0120050EA 359 EUR
ENZOBMLHP0120050
12(S)-Hydroperoxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HPETE) ≥98%
12(S)-Hydroperoxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HPETE)

Primary metabolite of arachidonic acid via the 12-lipoxygenase pathway. Inhibits thromboxane synthase. Inhibits platelet aggregation via activation of guanylate cyclase (100% at 5µM). Stimulates leukotriene biosynthesis. Potent and selective inhibitor of Ca2+-calmodulin-dependent protein kinase II (IC50=0.7µM). Endogenous TRPV1 agonist (Ki=0.35µM).


  • Thromboxane synthase inhibitor


Liquid.


Soluble in DMSO or dimethyl formamide; slightly soluble in PBS, pH 7.2 (0.8mg/ml) or 0.1M Na2CO3 (2mg/ml).

Formel: C₂₀H₃₂O₄
Molecular Weight: 336,5 g/mol
Lagertemperatur: Ultra-Tiefkühlschrank (–80°C)
CAS-Nummer: 71774-10-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=428867265ebd26c4753526856b0414bf8dd96a58&vid=582ee60bb6249d23094f4aaa36e32d25cc1ab74d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR