Blocking peptide for Heme oxygenase 1 pAb (Prod. No. BmL-HC3001) Blocking Peptide
Lieferant: ENZO LIFE SCIENCES
BMLHP93010100
ENZOBMLHP93010100EA 101 EUR
ENZOBMLHP93010100
Blocking peptide for Heme oxygenase 1 pAb (Prod. No. BmL-HC3001) Blocking Peptide
Proteine und Peptide
Synthetic peptide corresponding to amino acid residues [12-25] + Cys-NH2 of human heme oxygenase-1.
Complementary peptide to rabbit polyclonal antibody (BML-HC3001) for use in control experiments.
Preincubation of antibody with this peptide abolishes the antibody reactivity with HO-1 protein.