5-Ketoeicosa-6E,8Z,11Z,14E-tetraenoic acid ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

BMLK0050050
ENZOBMLK0050050EA 228 EUR
ENZOBMLK0050050
5-Ketoeicosa-6E,8Z,11Z,14E-tetraenoic acid ≥95% (durch HPLC)
5-Ketoeicosa-6E,8Z,11Z,14E-tetraenoic acid

R527 ligand.


Stimulates neutrophil degranulation and Ca²⁺ mobilisation by a mechanism independent of the LTB4 receptor. Has been shown to act as an agonist at a G protein-coupled receptor (R527). Induces the infiltration of granulocytes into human skin.

Formel: C₂₀H₃₀O₃
Lagertemperatur: Ultra-Tiefkühlschrank (–80°C)
CAS-Nummer: 106154-18-1
UN: 1170
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c196a014f645015240ba32fde8cc8b7dc31decbb&vid=daa148d48579e1a003efd20e94b25b6b9e5b4580&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Concentration 50 µg/ml
Formulation Oil dissolved in ethanol
Purity ≥95% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR